Product Details
- SNP ID
-
rs4381495
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.14:36474001 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTTTAGCAATTAAACAAAAGTATTT[C/T]TAAATCTCTCTATATTATGTTTTTC
- Phenotype
-
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
SFTA3
PubMed Links
Gene Details
- Gene
- SFTA3
- Gene Name
- surfactant associated 3
View Full Product Details