Product Details
- SNP ID
-
rs4810646
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:47507067 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTGGTGGCTATGGCAGCTGAGTTGG[A/C]GTGGTAGATGGATAGATGGGAAAGG
- Phenotype
-
MIM: 601937
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
NCOA3
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs115211941] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- NCOA3
- Gene Name
- nuclear receptor coactivator 3
View Full Product Details