Product Details

SNP ID
rs4976693
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.5:176814919 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCAGCAACCATCCCATCCCTGTCC[T/C]TGAACACGGACTGCCATGCTCCAGG
Phenotype
MIM: 607869
Polymorphism
T/C, Transition Substitution
Allele Nomenclature
Literature Links
UNC5A PubMed Links

Gene Details

Gene
UNC5A
Gene Name
unc-5 netrin receptor A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_133369.2 Intron NP_588610.2

View Full Product Details