Product Details

SNP ID
rs4746263
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:75103598 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCATCCACAGACCTTTGTCATACAC[A/G]CATACCTTGATACTACTCCCCGCTT
Phenotype
MIM: 613191 MIM: 611575
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DUSP13 PubMed Links

Gene Details

Gene
DUSP13
Gene Name
dual specificity phosphatase 13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001007271.1 Intron NP_001007272.1
NM_001007272.1 Intron NP_001007273.1
NM_001007273.1 Intron NP_001007274.1
NM_001320842.1 Intron NP_001307771.1
NM_001320843.1 Intron NP_001307772.1
NM_016364.3 Intron NP_057448.3
XM_005269883.3 Intron XP_005269940.1
XM_005269884.4 Intron XP_005269941.3
XM_005269887.1 Intron XP_005269944.1
XM_005269890.1 Intron XP_005269947.1
XM_011539853.1 Intron XP_011538155.1
XM_011539854.2 Intron XP_011538156.1
XM_011539855.1 Intron XP_011538157.1
XM_011539856.2 Intron XP_011538158.1
XM_017016313.1 Intron XP_016871802.1
XM_017016314.1 Intron XP_016871803.1
Gene
SAMD8
Gene Name
sterile alpha motif domain containing 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001174156.1 Intron NP_001167627.1
NM_144660.2 Intron NP_653261.1
XM_005269541.4 Intron XP_005269598.1
XM_011539311.1 Intron XP_011537613.1
XM_011539312.2 Intron XP_011537614.1
XM_017015738.1 Intron XP_016871227.1
XM_017015739.1 Intron XP_016871228.1

View Full Product Details