Product Details
- SNP ID
-
rs4653460
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
15
- Location
-
Chr.1:226656453 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TATCAGGGCCCTGACATTCCTCTCC[C/T]GGAAGCCACAGCTGTGCTGGGACAA
- Phenotype
-
MIM: 147522
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
ITPKB
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs78700535] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ITPKB
- Gene Name
- inositol-trisphosphate 3-kinase B
- Gene
- ITPKB-IT1
- Gene Name
- ITPKB intronic transcript 1
There are no transcripts associated with this gene.
View Full Product Details