Product Details

SNP ID
rs4976745
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:178184675 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AACACAAAAGGTCAACATATATAAA[A/T]CATGACAAGTGTACATCTTATTTTT
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
GMCL1P1 PubMed Links

Gene Details

Gene
GMCL1P1
Gene Name
germ cell-less, spermatogenesis associated 1 pseudogene 1
There are no transcripts associated with this gene.

View Full Product Details