Product Details

SNP ID
rs4860991
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:41360489 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATTCCCTCTATCGGGCCGGGTGTGA[A/G]CGGGAGGGAGACACGCAGCGCTCGC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LIMCH1 PubMed Links

Gene Details

Gene
LIMCH1
Gene Name
LIM and calponin homology domains 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001112717.2 Intron NP_001106188.1
NM_001112718.2 Intron NP_001106189.1
NM_001112719.2 Intron NP_001106190.1
NM_001112720.2 Intron NP_001106191.1
NM_001289122.1 Intron NP_001276051.1
NM_001289124.1 Intron NP_001276053.1
NM_014988.3 Intron NP_055803.2
XM_005248057.1 Intron XP_005248114.1
XM_005248058.1 Intron XP_005248115.1
XM_005248060.1 Intron XP_005248117.1
XM_005248061.1 Intron XP_005248118.1
XM_005248067.2 Intron XP_005248124.1
XM_005248072.1 Intron XP_005248129.1
XM_005248074.1 Intron XP_005248131.1
XM_005248075.1 Intron XP_005248132.1
XM_006713996.1 Intron XP_006714059.1
XM_011513642.1 Intron XP_011511944.1
XM_011513643.1 Intron XP_011511945.1
XM_011513644.1 Intron XP_011511946.1
XM_011513645.1 Intron XP_011511947.1
XM_011513646.1 Intron XP_011511948.1
XM_011513648.1 Intron XP_011511950.1
XM_011513649.1 Intron XP_011511951.1
XM_011513651.2 Intron XP_011511953.1
XM_011513653.1 Intron XP_011511955.1
XM_011513654.1 Intron XP_011511956.1
XM_011513655.1 Intron XP_011511957.1
XM_011513656.1 Intron XP_011511958.1
XM_011513657.1 Intron XP_011511959.1
XM_011513658.2 Intron XP_011511960.1
XM_017007894.1 Intron XP_016863383.1
XM_017007895.1 Intron XP_016863384.1
XM_017007896.1 Intron XP_016863385.1
XM_017007897.1 Intron XP_016863386.1
XM_017007898.1 Intron XP_016863387.1
XM_017007899.1 Intron XP_016863388.1
XM_017007900.1 Intron XP_016863389.1
XM_017007901.1 Intron XP_016863390.1
XM_017007902.1 Intron XP_016863391.1
XM_017007903.1 Intron XP_016863392.1
XM_017007904.1 Intron XP_016863393.1
XM_017007905.1 Intron XP_016863394.1

View Full Product Details