Product Details
- SNP ID
-
rs4917407
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:103968116 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- ATGGAGTTTAAAGAGCGATTGGAAA[A/G]GGAGACAGGGTTATAAAGTCTACTC
- Phenotype
-
MIM: 616563
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
SLK
PubMed Links
Gene Details
- Gene
- SLK
- Gene Name
- STE20 like kinase
View Full Product Details