Product Details

SNP ID
rs4703484
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:34907064 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTGAGACACTTACTAGTTATCAGGT[A/G]CTGACCCTGGCCACCTCCCAGAATT
Phenotype
MIM: 603153 MIM: 616344
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BRIX1 PubMed Links

Gene Details

Gene
BRIX1
Gene Name
BRX1, biogenesis of ribosomes
There are no transcripts associated with this gene.

Gene
RAD1
Gene Name
RAD1 checkpoint DNA exonuclease
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002853.3 2879 UTR 3 NP_002844.1
Gene
TTC23L
Gene Name
tetratricopeptide repeat domain 23 like
There are no transcripts associated with this gene.

View Full Product Details