Product Details

SNP ID
rs4813786
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:6007449 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTAATCTGAGATTAGCTCTCCTAAC[A/G]TTAACTTTGAAACTTTTACGGATTT
Phenotype
MIM: 608188 MIM: 608187
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CRLS1 PubMed Links

Gene Details

Gene
CRLS1
Gene Name
cardiolipin synthase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001127458.1 Intron NP_001120930.1
NM_001323561.1 Intron NP_001310490.1
NM_001323562.1 Intron NP_001310491.1
NM_001323563.1 Intron NP_001310492.1
NM_001323564.1 Intron NP_001310493.1
NM_019095.5 Intron NP_061968.1
XM_005260738.4 Intron XP_005260795.1
XM_011529263.1 Intron XP_011527565.1
Gene
MCM8
Gene Name
minichromosome maintenance 8 homologous recombination repair factor
There are no transcripts associated with this gene.

Gene
MCM8-AS1
Gene Name
MCM8 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details