Product Details

SNP ID
rs4742906
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:104094797 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAGTCAAGCACAATTCGGGGATGCA[A/G]TGACGGACGTAGGTGAAGACACTGC
Phenotype
MIM: 605576
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SMC2 PubMed Links

Gene Details

Gene
SMC2
Gene Name
structural maintenance of chromosomes 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001042550.1 227 UTR 5 NP_001036015.1
NM_001042551.1 227 Intron NP_001036016.1
NM_001265602.1 227 Intron NP_001252531.1
NM_006444.2 227 Intron NP_006435.2
XM_006716933.3 227 Intron XP_006716996.1
XM_011518148.2 227 Intron XP_011516450.1
XM_011518149.2 227 Intron XP_011516451.1
XM_011518150.2 227 Intron XP_011516452.1
XM_011518151.2 227 Intron XP_011516453.1
XM_011518152.2 227 Intron XP_011516454.1
XM_011518153.1 227 Intron XP_011516455.1
XM_017014206.1 227 UTR 5 XP_016869695.1
XM_017014207.1 227 Intron XP_016869696.1
XM_017014208.1 227 UTR 5 XP_016869697.1
XM_017014209.1 227 UTR 5 XP_016869698.1
XM_017014210.1 227 Intron XP_016869699.1
XM_017014211.1 227 Intron XP_016869700.1
XM_017014212.1 227 Intron XP_016869701.1
Gene
SMC2-AS1
Gene Name
SMC2 antisense RNA 1 (head to head)
There are no transcripts associated with this gene.

View Full Product Details