Product Details
- SNP ID
-
hCV27926999
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:115120717 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGTAGGGCTTGAAACAATCTATCTA[C/T]TTAAATGTATTGTGAATGGAGCATA
- Phenotype
-
MIM: 612869
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ATRNL1
PubMed Links
Gene Details
- Gene
- ATRNL1
- Gene Name
- attractin like 1
View Full Product Details