Product Details

SNP ID
rs4550486
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:66877760 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTTTCACCTCCCAGGTCTTCCTTC[C/T]TGAAAGATGAATGTAACTATGCCTT
Phenotype
MIM: 606405
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CACNG5 PubMed Links

Gene Details

Gene
CACNG5
Gene Name
calcium voltage-gated channel auxiliary subunit gamma 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145811.2 Intron NP_665810.1

View Full Product Details