Product Details
- SNP ID
-
rs4888136
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:81315886 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TCCTTGGGTTGGTAGTCCGAACCTC[C/T]GGGCGAAATTCTCCTTGCCAAACGT
- Phenotype
-
MIM: 605379
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
GAN
PubMed Links
Gene Details
- Gene
- GAN
- Gene Name
- gigaxonin
View Full Product Details