Product Details

SNP ID
rs4323883
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:125496794 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGATGCTGGGTGGGAGCTGGATCCA[C/G]CTGAAAAATCTGAGTGAGAAGGTTT
Phenotype
MIM: 604825
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
FEZ1 PubMed Links

Gene Details

Gene
FEZ1
Gene Name
fasciculation and elongation protein zeta 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005103.4 689 Intron NP_005094.1
NM_022549.3 689 Intron NP_072043.1
Gene
LOC403312
Gene Name
putative uncharacterized protein MGC39545
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001301851.1 689 Missense Mutation CCT,GCT P192A NP_001288780.1

View Full Product Details