Product Details
- SNP ID
-
rs4610427
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:9038440 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GAAAACCCCATTGAAGTGTAAAGGA[G/T]ACCCGGACCCTAAAGAAAGCTGATG
- Phenotype
-
MIM: 609297
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
SEMA5A
PubMed Links
Gene Details
- Gene
- SEMA5A
- Gene Name
- semaphorin 5A
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_003966.2 |
8394 |
UTR 3 |
|
|
NP_003957.2 |
View Full Product Details