Product Details

SNP ID
rs4580247
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:82247962 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCCCCACTCCGGCATGTTCCTGGC[A/T]AGCCAGCTACACCCAGCCCCGCTCA
Phenotype
MIM: 600864 MIM: 603877
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
CSNK1D PubMed Links

Gene Details

Gene
CSNK1D
Gene Name
casein kinase 1 delta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001893.4 Intron NP_001884.2
NM_139062.2 Intron NP_620693.1
Gene
SLC16A3
Gene Name
solute carrier family 16 member 3
There are no transcripts associated with this gene.

View Full Product Details