Product Details
- SNP ID
-
hCV27933310
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:120711283 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGATGGGTTCTTCCTCAAAACACAT[A/C]GCCGTGGCATTTCCTGCTCATTTAA
- Phenotype
-
MIM: 613802
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
MLEC
PubMed Links
Gene Details
- Gene
- MLEC
- Gene Name
- malectin
There are no transcripts associated with this gene.
- Gene
- UNC119B
- Gene Name
- unc-119 lipid binding chaperone B
View Full Product Details