Product Details

SNP ID
rs4803880
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.19:45997036 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACCTATTGGGTTCCTCCTCCTCCCT[C/T]CCCTCAACTCCCATCACTTCTCTCA
Phenotype
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
CCDC61 PubMed Links
Additional Information
For this assay, SNP(s) [rs143285601] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
CCDC61
Gene Name
coiled-coil domain containing 61
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001267723.1 Intron NP_001254652.1
XM_005259191.3 Intron XP_005259248.1
XM_005259192.4 Intron XP_005259249.1
XM_011527257.2 Intron XP_011525559.1

View Full Product Details