Product Details

SNP ID
rs4528960
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:84959226 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGTGACCTACCCACTCCTTGCAGCC[C/T]TCGCCCGCACCTTCTCCAACACCCC
Phenotype
MIM: 609938
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CADM2 PubMed Links

Gene Details

Gene
CADM2
Gene Name
cell adhesion molecule 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001167674.1 245 UTR 5 NP_001161146.1
NM_001167675.1 245 UTR 5 NP_001161147.1
NM_001256502.1 245 Intron NP_001243431.1
NM_001256503.1 245 Intron NP_001243432.1
NM_001256504.1 245 Intron NP_001243433.1
NM_001256505.1 245 Intron NP_001243434.1
NM_153184.3 245 Intron NP_694854.2

View Full Product Details