Product Details

SNP ID
rs4436441
Assay Type
Functionally tested
NCBI dbSNP Submissions
71
Location
Chr.1:202118808 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGAAAGCCTTCCATCTCTGCCTGG[A/C]ATGCTTCTGAATAGGGGAGATGGCT
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
GPR37L1 PubMed Links

Gene Details

Gene
GPR37L1
Gene Name
G protein-coupled receptor 37 like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004767.3 140 Intron NP_004758.3
XM_011510158.2 140 UTR 5 XP_011508460.1

View Full Product Details