Product Details

SNP ID
rs4339951
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:92296168 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGATTACAGGGGTGAGCCACCACT[C/T]CTGACCATATGGATATATATTCTTT
Phenotype
MIM: 610606 MIM: 610637
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CPEB3 PubMed Links

Gene Details

Gene
CPEB3
Gene Name
cytoplasmic polyadenylation element binding protein 3
There are no transcripts associated with this gene.

Gene
MARCH5
Gene Name
membrane associated ring-CH-type finger 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017824.4 Intron NP_060294.1

View Full Product Details