Product Details

SNP ID
rs4720027
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:31058504 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCCCTTACAGTCCAGGTATCTGTA[A/T]CACATCCCATTCCTGCCAGGCAAGC
Phenotype
MIM: 102981
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
ADCYAP1R1 PubMed Links

Gene Details

Gene
ADCYAP1R1
Gene Name
ADCYAP receptor type I
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001118.4 Intron NP_001109.2
NM_001199635.1 Intron NP_001186564.1
NM_001199636.1 Intron NP_001186565.1
NM_001199637.1 Intron NP_001186566.1

View Full Product Details