Product Details

SNP ID
rs3923811
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:16415935 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGCCTAGTGGGCTCCTGGCTTCTA[C/T]AGCCTGGAGCCAGGGTCTGTGTGGG
Phenotype
MIM: 606676
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TRPV2 PubMed Links

Gene Details

Gene
TRPV2
Gene Name
transient receptor potential cation channel subfamily V member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016113.4 Intron NP_057197.2
XM_005256676.1 Intron XP_005256733.1
XM_005256677.1 Intron XP_005256734.1
XM_005256678.4 Intron XP_005256735.1
XM_006721541.3 Intron XP_006721604.1
XM_006721543.3 Intron XP_006721606.1
XM_011523922.1 Intron XP_011522224.1
XM_011523923.1 Intron XP_011522225.1
XM_011523925.2 Intron XP_011522227.1
XM_017024730.1 Intron XP_016880219.1
XM_017024731.1 Intron XP_016880220.1
XM_017024732.1 Intron XP_016880221.1

View Full Product Details