Product Details

SNP ID
rs3943363
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:56743540 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTACTCTTTCACTTAATCATTGATA[C/T]AACTGTGAGACAGGTAGAGTGCATG
Phenotype
MIM: 606623 MIM: 176635
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HSD17B6 PubMed Links

Gene Details

Gene
HSD17B6
Gene Name
hydroxysteroid 17-beta dehydrogenase 6
There are no transcripts associated with this gene.

Gene
PRIM1
Gene Name
primase (DNA) subunit 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000946.2 Intron NP_000937.1

View Full Product Details