Product Details
- SNP ID
-
rs4938357
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:117213324 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGATTAAATTGCAAGGGAAAGGGCT[A/G]GGGAAGACCTGGGAAGAGGCGAGGG
- Phenotype
-
MIM: 604872
MIM: 600818
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
PCSK7
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs76553074] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- PCSK7
- Gene Name
- proprotein convertase subtilisin/kexin type 7
- Gene
- TAGLN
- Gene Name
- transgelin
There are no transcripts associated with this gene.
View Full Product Details