Product Details
- SNP ID
-
rs4839027
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
30
- Location
-
Chr.1:110481567 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TTTGCACTCTTGCCATCACATCACG[C/T]GGTAATCTACAGGTGCTCAGGTTTG
- Phenotype
-
MIM: 118943
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CYMP
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs116810605] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CYMP
- Gene Name
- chymosin pseudogene
There are no transcripts associated with this gene.
- Gene
- LOC440602
- Gene Name
- uncharacterized LOC440602
There are no transcripts associated with this gene.
View Full Product Details