Product Details

SNP ID
hCV27941044
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:61549851 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAATGAACTTTTTGTATTGTTACA[A/T]TAAAATCTATTAGGTTATATTCTAG
Phenotype
MIM: 610487
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
KHDRBS2 PubMed Links

Gene Details

Gene
KHDRBS2
Gene Name
KH RNA binding domain containing, signal transduction associated 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152688.3 Intron NP_689901.2

View Full Product Details