Product Details

SNP ID
rs4665172
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:160276081 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATCATTTAATGCCTCCAGTAGGGT[C/G]TAGGGCTGCACCTCATAAGCAAACA
Phenotype
MIM: 602310
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LOC100505984 PubMed Links

Gene Details

Gene
LOC100505984
Gene Name
uncharacterized LOC100505984
There are no transcripts associated with this gene.

Gene
RBMS1
Gene Name
RNA binding motif single stranded interacting protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002897.4 Intron NP_002888.1
NM_016836.3 Intron NP_058520.1

View Full Product Details