Product Details

SNP ID
rs906351553
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:154836412 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAATCTTATGGGGGTGGAGGGCAA[A/G]AAGGGGGATCCTATTGTGTGGTGAT
Phenotype
MIM: 300841
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
F8 PubMed Links

Gene Details

Gene
F8
Gene Name
coagulation factor VIII
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000132.3 1981 UTR 3 NP_000123.1
NM_019863.2 1981 UTR 3 NP_063916.1
Gene
SMIM9
Gene Name
small integral membrane protein 9
There are no transcripts associated with this gene.

View Full Product Details