Product Details

SNP ID
rs4232893
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:30412430 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCCAAAATGAAGGAAGACTGTGACC[G/A]CAAAAAGGGGCCAGGGGAGGGAGAC
Phenotype
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
ZNF771 PubMed Links

Gene Details

Gene
ZNF771
Gene Name
zinc finger protein 771
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142305.1 Intron NP_001135777.1
NM_016643.3 Intron NP_057727.2

View Full Product Details