Product Details

SNP ID
rs4808844
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:18701156 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCACTGCCGGCTCAGCCGGGGCAGA[A/G]GGAGGAGCCTGAAGGTCCACCATGG
Phenotype
MIM: 607536
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CRTC1 PubMed Links

Gene Details

Gene
CRTC1
Gene Name
CREB regulated transcription coactivator 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001098482.1 Intron NP_001091952.1
NM_015321.2 Intron NP_056136.2
XM_005259833.3 Intron XP_005259890.1
XM_005259834.1 Intron XP_005259891.1
XM_005259835.3 Intron XP_005259892.1
XM_005259836.3 Intron XP_005259893.1
XM_006722710.3 Intron XP_006722773.1
XM_011527842.2 Intron XP_011526144.1
XM_017026536.1 Intron XP_016882025.1
XM_017026537.1 Intron XP_016882026.1
XM_017026538.1 Intron XP_016882027.1

View Full Product Details