Product Details

SNP ID
rs4380916
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:124307930 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGTAAAGAAGAGAAGTGAGCACCA[C/G]GGTTTAAGGCAGAAAGGGACAGGCT
Phenotype
MIM: 616609
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
TMEM65 PubMed Links

Gene Details

Gene
TMEM65
Gene Name
transmembrane protein 65
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_194291.2 Intron NP_919267.2
XM_011516847.2 Intron XP_011515149.1
XM_017013104.1 Intron XP_016868593.1
XM_017013105.1 Intron XP_016868594.1

View Full Product Details