Product Details

SNP ID
rs4996209
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:28907402 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCTCTGTTGTTAGTCATGCACTAA[G/T]TTTTTCATATGATGTATGGCTCTAT
Phenotype
MIM: 602165
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
TRIM27 PubMed Links

Gene Details

Gene
TRIM27
Gene Name
tripartite motif containing 27
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006510.4 Intron NP_006501.1

View Full Product Details