Product Details

SNP ID
rs4720675
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:6465716 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCTGTGGCTACTGAAAATTATGTAG[A/G]TTTCTAGTGACATGAAAAATGTGAG
Phenotype
MIM: 609024
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KDELR2 PubMed Links

Gene Details

Gene
KDELR2
Gene Name
KDEL endoplasmic reticulum protein retention receptor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001100603.1 Intron NP_001094073.1
NM_006854.3 Intron NP_006845.1

View Full Product Details