Product Details

SNP ID
rs4910187
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:10755484 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAAATTTTTCTTTTGGTTGTTGTTA[C/T]TGGCACAGAAGACTTACATTTCAGG
Phenotype
MIM: 609366
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CTR9 PubMed Links

Gene Details

Gene
CTR9
Gene Name
CTR9 homolog, Paf1/RNA polymerase II complex component
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014633.4 Intron NP_055448.1

View Full Product Details