Product Details
- SNP ID
-
rs4778531
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.15:23443797 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CTTACCCTGTCTGCCTTCTTGTGCC[A/G]TGTGGACACAGCAGAGAGAGCCCGC
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
GOLGA6L2
PubMed Links
Gene Details
- Gene
- GOLGA6L2
- Gene Name
- golgin A6 family-like 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001304388.1 |
633 |
Missense Mutation |
CGG,TGG |
R191W |
NP_001291317.1 |
View Full Product Details