Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001166244.1 | 3157 | UTR 3 | NP_001159716.1 | ||
NM_001166245.1 | 3157 | UTR 3 | NP_001159717.1 | ||
NM_001166246.1 | 3157 | UTR 3 | NP_001159718.1 | ||
NM_021828.4 | 3157 | UTR 3 | NP_068600.4 | ||
XM_006717937.2 | 3157 | UTR 3 | XP_006718000.1 | ||
XM_011540029.1 | 3157 | UTR 3 | XP_011538331.1 | ||
XM_011540030.1 | 3157 | UTR 3 | XP_011538332.1 | ||
XM_011540031.2 | 3157 | UTR 3 | XP_011538333.1 | ||
XM_011540033.2 | 3157 | UTR 3 | XP_011538335.1 | ||
XM_017016495.1 | 3157 | Intron | XP_016871984.1 | ||
XM_017016496.1 | 3157 | Intron | XP_016871985.1 | ||
XM_017016497.1 | 3157 | UTR 3 | XP_016871986.1 | ||
XM_017016498.1 | 3157 | UTR 3 | XP_016871987.1 |