Product Details

SNP ID
rs4925827
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:144493267 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGCTTTGAGGAGTGGAATGAACCC[A/C]CCGGGTTATCTGTTTAGAACTGGAA
Phenotype
MIM: 138200 MIM: 609172
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
GPT PubMed Links

Gene Details

Gene
GPT
Gene Name
glutamic--pyruvic transaminase
There are no transcripts associated with this gene.

Gene
LOC101928953
Gene Name
uncharacterized LOC101928953
There are no transcripts associated with this gene.

Gene
PPP1R16A
Gene Name
protein phosphatase 1 regulatory subunit 16A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_005272350.2 Intron XP_005272407.1
XM_011517349.2 Intron XP_011515651.1
XM_017013912.1 Intron XP_016869401.1
XM_017013913.1 Intron XP_016869402.1
XM_017013914.1 Intron XP_016869403.1
XM_017013915.1 Intron XP_016869404.1

View Full Product Details