Product Details

SNP ID
rs4461125
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:41724428 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGATAGAGGAGGCAGGGGTTGGGGG[C/T]AGGGGAAGCAAGCGGGCAGAGATGG
Phenotype
MIM: 137250 MIM: 600947
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GAST PubMed Links

Gene Details

Gene
GAST
Gene Name
gastrin
There are no transcripts associated with this gene.

Gene
HAP1
Gene Name
huntingtin associated protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001079870.1 2070 UTR 3 NP_001073339.1
NM_001079871.1 2070 UTR 3 NP_001073340.1
NM_177977.2 2070 UTR 3 NP_817084.2

View Full Product Details