Product Details

SNP ID
rs4596174
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:183644725 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTAATTGATACAAAGCTTAATCAAT[A/T]TTATATTAATACTTAACTCACTGTT
Phenotype
MIM: 611295
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
KLHL24 PubMed Links

Gene Details

Gene
KLHL24
Gene Name
kelch like family member 24
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017644.3 Intron NP_060114.2

View Full Product Details