Product Details

SNP ID
rs4448892
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:81196722 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGTGGCTCTGCACTGGGTTTGCCT[C/T]CTGGGTGTTGCCGGCCAGCCCAGCA
Phenotype
MIM: 603035
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IL16 PubMed Links

Gene Details

Gene
IL16
Gene Name
interleukin 16
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001172128.1 Intron NP_001165599.1
NM_004513.5 Intron NP_004504.3
NM_172217.3 Intron NP_757366.2
XM_005254342.3 Intron XP_005254399.1
XM_005254346.4 Intron XP_005254403.1
XM_011521519.2 Intron XP_011519821.1
XM_011521520.2 Intron XP_011519822.1
XM_017022140.1 Intron XP_016877629.1
XM_017022141.1 Intron XP_016877630.1
XM_017022142.1 Intron XP_016877631.1
XM_017022143.1 Intron XP_016877632.1
XM_017022144.1 Intron XP_016877633.1
XM_017022145.1 Intron XP_016877634.1
XM_017022146.1 Intron XP_016877635.1

View Full Product Details