Product Details

SNP ID
hCV27960677
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:3464843 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCCTCAAGAGGGTGTGCTCCCCCG[C/G]CCGGTGAGTGCCTGAGTGGAGTGAC
Phenotype
MIM: 610285
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
DOK7 PubMed Links

Gene Details

Gene
DOK7
Gene Name
docking protein 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001164673.1 Intron NP_001158145.1
NM_001256896.1 Intron NP_001243825.1
NM_001301071.1 Intron NP_001288000.1
NM_173660.4 Intron NP_775931.3

View Full Product Details