Product Details

SNP ID
rs4766042
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:3083861 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAGGATGCTGACATTTTCTAGTTGT[C/T]GTGCCCTGACTCCTGGCTTTGATTT
Phenotype
MIM: 613137
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TSPAN9 PubMed Links

Gene Details

Gene
TSPAN9
Gene Name
tetraspanin 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001168320.1 Intron NP_001161792.1
NM_006675.4 Intron NP_006666.1
XM_011520912.2 Intron XP_011519214.1
XM_011520913.2 Intron XP_011519215.1

View Full Product Details