Product Details
- SNP ID
-
rs4869249
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:95722240 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CAGGAGATGGGGAAGGCTTTTAGTA[C/G]AGTTAAAACATGAGCCAAATTATAG
- Phenotype
-
MIM: 607353
MIM: 608039
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
RHOBTB3
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs146348608,rs79014317] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- RHOBTB3
- Gene Name
- Rho related BTB domain containing 3
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_014899.3 |
|
Intron |
|
|
NP_055714.3 |
- Gene
- SPATA9
- Gene Name
- spermatogenesis associated 9
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_031952.3 |
|
Intron |
|
|
NP_114158.2 |
View Full Product Details