Product Details
- SNP ID
-
rs4146950
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:146177347 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACCTGGGTTATTCTCTCCTTTTAAA[A/G]GTGTACTAACAGATTTTAATCATCG
- Phenotype
-
MIM: 607286
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
LSM6
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs74537062] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- LSM6
- Gene Name
- LSM6 homolog, U6 small nuclear RNA and mRNA degradation associated
View Full Product Details