Product Details
- SNP ID
-
rs4870855
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.8:123503614 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- ATGTTCTCACTCGTATGTGGGAGCT[A/G]TGAAAGTAGATTTCATGAAGACAGA
- Phenotype
-
MIM: 606604
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
FBXO32
PubMed Links
Gene Details
- Gene
- FBXO32
- Gene Name
- F-box protein 32
View Full Product Details