Product Details

SNP ID
rs4794037
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:49291940 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAAAATCTTGAATTTATTCATCATC[A/G]AACATAACCAATTCTCACAATTTTG
Phenotype
MIM: 613907
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MIR6129 PubMed Links

Gene Details

Gene
MIR6129
Gene Name
microRNA 6129
There are no transcripts associated with this gene.

Gene
ZNF652
Gene Name
zinc finger protein 652
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145365.1 8613 UTR 3 NP_001138837.1
NM_014897.2 8613 UTR 3 NP_055712.1
XM_005257166.2 8613 UTR 3 XP_005257223.1

View Full Product Details