Product Details

SNP ID
rs892041887
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:55009265 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTCATGAGCTCAAAGTGTACAGGA[C/T]GGCGACAGAAATTGCAGGCAGCCAC
Phenotype
MIM: 301300 MIM: 300773
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ALAS2 PubMed Links

Gene Details

Gene
ALAS2
Gene Name
5'-aminolevulinate synthase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000032.4 1813 Missense Mutation CAT,CGT H560R NP_000023.2
NM_001037967.3 1813 Missense Mutation CAT,CGT H523R NP_001033056.1
NM_001037968.3 1813 Missense Mutation CAT,CGT H547R NP_001033057.1
Gene
APEX2
Gene Name
apurinic/apyrimidinic endodeoxyribonuclease 2
There are no transcripts associated with this gene.

View Full Product Details