Product Details

SNP ID
rs4596138
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.3:53498867 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCATTTTGCAGCAGATGTCCCTTCT[A/C]TAGAGTGAGTTATGGTGTCTCTCAC
Phenotype
MIM: 114206
Polymorphism
A/C, Transversion Substitution
Allele Nomenclature
Literature Links
CACNA1D PubMed Links

Gene Details

Gene
CACNA1D
Gene Name
calcium voltage-gated channel subunit alpha1 D
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000720.3 Intron NP_000711.1
NM_001128839.2 Intron NP_001122311.1
NM_001128840.2 Intron NP_001122312.1

View Full Product Details